Assay Details
| Gene Symbol: ZNF483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011518300.2 | XP_011516602.1 | 6 | 9607 | 112 | |
| XM_017014337.1 | XP_016869826.1 | 7 | 8580 | 112 | ||
| XM_017014338.1 | XP_016869827.1 | 7 | 8466 | 112 | ||
| GenBank mRNA | AK054857.1 | - | 1 | 249 | 112 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | KIAA0368 | KIAA0368 | ECM29 | -64.1kb | Anti-sense | Chr. 9 - 111360693 - 111484745 on NCBI Build GRCh38 |
| Downstream |
leucine rich repeat containing 37 member A5, pseudogene |
LRRC37A5P | C9orf29 | 51.9kb | Anti-sense | Chr. 9 - 111602831 - 111613553 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ZNF483
Genomic Map