Assay Details
| Gene Symbol: LOC105376090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK054896.1 | - | 1 | 859 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | osteoclast stimulating factor 1 | OSTF1 |
OSF, SH3P2, bA235O14.1 |
-18.9kb | Sense | Chr. 9 - 75088480 - 75147201 on NCBI Build GRCh38 |
| Downstream | proprotein convertase subtilisin/kexin type 5 | PCSK5 |
PC5, PC6, PC6A, SPC6 |
721.3kb | Sense | Chr. 9 - 75890601 - 76362339 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC105376090
Genomic Map