Assay Details
| Gene Symbol: LYPD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182573.2 | NP_872379.2 | 1 - 2 | 118 | 80 | |
| GenBank mRNA | AK055031.1 | - | 1 - 2 | 96 | 80 | |
| DA286806.1 | - | 1 - 2 | 118 | 80 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001596.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK055031.1,NM_182573.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19021864.hg.1 | ENST00000377950.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021864.hg.1 | ENST00000414615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021864.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021864.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021864.hg.1 | TC19001596.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021867.hg.1 | ENST00000414615.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021867.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021867.hg.1 | TC19001596.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000594013.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000594013.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000594013.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000594013.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000594013.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000594013.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000594013.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000594013.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000594013.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000594049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000594049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000594049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000594049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000594049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000594049.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000595666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000595666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000595666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000595666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000595666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000601224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000601224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000601224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | ENST00000601224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_001288763.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_001288763.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_001288763.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_001288763.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_001288763.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_001288763.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_001288763.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_001288763.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_001288763.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_001288763.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_001288763.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011970.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206610.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206610.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206610.hg.1 | TC1900011970.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900206610.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206610.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206603.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206603.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206603.hg.1 | ENST00000594013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206603.hg.1 | ENST00000594049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206603.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206603.hg.1 | NM_001288763.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206603.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206603.hg.1 | TC1900011970.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900206603.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206603.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206603.hg.1 | ENST00000594013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206603.hg.1 | ENST00000594049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206603.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206603.hg.1 | NM_001288763.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206603.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000377950.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000594013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000594013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000594013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000594013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000594013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000594013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000594013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000594013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000594013.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000594049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000594049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000594049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000594049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000594049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000594049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000595666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000595666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000595666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000595666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000595666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000601224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000601224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000601224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | ENST00000601224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_001031749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_001288763.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_001288763.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_001288763.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_001288763.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_001288763.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_001288763.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_001288763.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_001288763.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_001288763.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_001288763.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_001288763.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011970.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206609.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206609.hg.1 | ENST00000594049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206609.hg.1 | NM_182573.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900206609.hg.1 | TC1900011970.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900206609.hg.1 | ENST00000414615.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206609.hg.1 | ENST00000594049.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900206609.hg.1 | NM_182573.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 120674 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 180917 | - | - |

LYPD5
Genomic Map