Assay Details
| Gene Symbol: QSER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076786.2 | NP_001070254.1 | 13 | 7396 | 103 | |
| XM_017018328.1 | XP_016873817.1 | 13 | 7748 | 103 | ||
| GenBank mRNA | AK055185.1 | - | 1 | 1851 | 103 | |
| AK095528.1 | - | 1 | 472 | 103 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

QSER1
Genomic Map