Assay Details
| Gene Symbol: LINC01529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104176.1 | 1 - 2 | 113 | 77 | ||
| GenBank mRNA | AK055260.1 | - | 1 - 2 | 89 | 77 | |
| DA464500.1 | - | 1 - 2 | 113 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3775281 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 30100 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3775281 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03446283 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P13031504 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P41186761 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03446283 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | ENST00000564335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | ENST00000564335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | ENST00000564335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | ENST00000567313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | ENST00000567313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | ENST00000567313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | ENST00000567313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | ENST00000567313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010498.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900185935.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185935.hg.1 | ENST00000567313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185935.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900185935.hg.1 | TC1900010498.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900185935.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185935.hg.1 | ENST00000567313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185935.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900185936.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185936.hg.1 | ENST00000564335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185936.hg.1 | ENST00000567313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185936.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900185936.hg.1 | TC1900010498.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900185936.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185936.hg.1 | ENST00000564335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185936.hg.1 | ENST00000567313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185936.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900185937.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185937.hg.1 | ENST00000564335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185937.hg.1 | ENST00000567313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185937.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900185937.hg.1 | TC1900010498.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900185937.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185937.hg.1 | ENST00000564335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185937.hg.1 | ENST00000567313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185937.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | ENST00000564335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | ENST00000564335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | ENST00000564335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | ENST00000567313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | ENST00000567313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | ENST00000567313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | ENST00000567313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | ENST00000567313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010498.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900185938.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185938.hg.1 | ENST00000567313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185938.hg.1 | NR_104176.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900185938.hg.1 | TC1900010498.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900185938.hg.1 | ENST00000433059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185938.hg.1 | ENST00000567313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185938.hg.1 | NR_104176.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 705058 | - | - |

LINC01529
Genomic Map