Assay Details
| Gene Symbol: RAB3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317915.1 | NP_001304844.1 | 5 | 7690 | 108 | |
| NM_138453.3 | NP_612462.1 | 5 | 7652 | 108 | ||
| GenBank mRNA | AB073648.1 | - | 1 | 822 | 108 | |
| AK055409.1 | - | 1 | 853 | 108 | ||
| AK094685.1 | - | 1 | 1699 | 108 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

RAB3C
Genomic Map