Assay Details
| Gene Symbol: DUSP5P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK055963.1 | - | 1 | 652 | 78 | |
| Gene Symbol: LOC105373132 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011544354.2 | XP_011542656.1 | 1 | 1982 | 78 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846289 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20916791 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846292 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846285 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846298 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20916808 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846283 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15189429 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846293 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20916798 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20916796 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846296 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846300 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20916803 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15189439 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846297 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846286 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20916810 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20916811 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15189442 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846281 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846280 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20916793 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846279 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20916780 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00846294 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 24385 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 2097 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 146681 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 14274 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 168062 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P367602 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P173790 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P367602 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P157846 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 2320 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 36090 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 7593 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P115799 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P115799 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 18368 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_14_P115799 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P15476352 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P367602 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P367602 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P367602 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P173790 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_32_P157846 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P115799 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P115799 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P35496088 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P115799 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P157846 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P367602 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 59161 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 101608 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 38318 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00269516 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P115799 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P10539403 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846285 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846277 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15189431 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15189426 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846298 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15189435 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846290 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846302 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846283 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15189437 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846297 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846292 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846303 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846281 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846300 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846284 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846279 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846295 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00846288 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_18_P10539403 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P115799 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_002834_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7910380 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7910382 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7910379 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7910381 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1553299_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017643.hg.1 | ENST00000390935.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017643.hg.1 | ENST00000390935.1 | Ensembl |
| Human Genome U95 Set | 74478_at | - | - |
| Human Genome U95 Set | 74478_PM_at | - | - |
| Human Genome U95 Set | 74476_PM_at | - | - |
| Human Genome U95 Set | 74476_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A041393 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A041386 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A041386 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A041393 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 227529 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1751093 | AK092260.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1751093 | NR_002834.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2051731 | AK092260.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2051731 | NR_002834.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1751093 | AK092260.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1751093 | NR_002834.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2051731 | AK092260.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2051731 | NR_002834.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2051731 | AK092260.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2051731 | NR_002834.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1751093 | AK092260.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1751093 | NR_002834.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2051731 | AK092260.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2051731 | NR_002834.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1751093 | AK092260.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1751093 | NR_002834.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1751093 | AK092260.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1751093 | NR_002834.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2051731 | AK092260.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2051731 | NR_002834.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2051731 | AK092260.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2051731 | NR_002834.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1751093 | AK092260.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1751093 | NR_002834.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2051731 | AK092260.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2051731 | NR_002834.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1751093 | AK092260.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1751093 | NR_002834.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2051731 | AK092260.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2051731 | NR_002834.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1751093 | AK092260.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1751093 | NR_002834.1 | human_transcriptome |

DUSP5P1
Genomic Map