Assay Details
| Gene Symbol: SRSF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK095580.1 | - | 1 | 856 | 82 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000613941.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000613941.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000613941.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000613941.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000613941.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620242.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000339436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000373715.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000477442.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000613941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000613941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000613941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000613941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000613941.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000614136.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620242.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | ENST00000620941.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NM_003017.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007842.hg.1 | NR_036610.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 104683 | - | - |

SRSF3
Genomic Map