Assay Details
| Gene Symbol: LOC93463 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135202.1 | 5 - 6 | 1095 | 77 | ||
| GenBank mRNA | AL122100.1 | - | 5 - 6 | 1099 | 77 | |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22113535 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986095 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986079 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22113557 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22113560 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22113551 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986113 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22113533 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986100 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22113580 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986067 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986122 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22113544 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986104 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986128 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986117 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22113542 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22113546 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986084 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986096 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986118 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22113534 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986126 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986098 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22113591 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22113555 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986129 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986109 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22113550 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22113574 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986088 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986114 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22113543 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986093 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986065 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986081 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986125 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22113563 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22113576 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986075 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22113531 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22113537 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986127 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22113540 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986091 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22113581 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986102 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986112 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986083 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986108 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986099 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22113582 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986070 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986124 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986107 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22113569 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P22113584 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986090 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P01986082 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3335266 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 18605 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 46568 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3335266 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P391731 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8059904 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8059903 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8059902 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 234861_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016181.hg.1 | ENST00000244820.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016181.hg.1 | ENST00000244820.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016181.hg.1 | ENST00000244820.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016181.hg.1 | ENST00000244820.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016181.hg.1 | ENST00000244820.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016181.hg.1 | ENST00000244820.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200016181.hg.1 | ENST00000244820.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016181.hg.1 | ENST00000244820.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016181.hg.1 | ENST00000244820.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016181.hg.1 | ENST00000244820.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016181.hg.1 | ENST00000244820.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016181.hg.1 | ENST00000244820.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016181.hg.1 | ENST00000244820.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200016181.hg.1 | ENST00000244820.2 | Ensembl |
| Human Genome U133 Plus 2.0 Array | 234861_at | - | - |
| Human Genome U133 Plus 2.0 Array | 234861_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234861_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234861_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234861_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234861_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234861_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234861_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234861_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234861_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234861_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234861_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 234861_at9 | - | - |
| Human Genome U133 Set | 234861_at | - | - |
| Human Genome U133 Set | 234861_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 124738 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 231700 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1777684 | AL122100.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1681611 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1681611 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1777684 | AL122100.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1777684 | AL122100.1 | human_transcriptome |

LOC93463
Genomic Map