Assay Details
Gene Symbol: BRWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007246.2 | NP_001007247.1 | 5 | 1901 | 131 | |
GenBank mRNA | AL109785.1 | - | 1 | 753 | 131 | |
AL109786.1 | - | 1 | 1168 | 131 | ||
AL162036.1 | - | 1 | 448 | 131 | ||
AL558272.3 | - | 1 | 358 | 131 | ||
BC031545.1 | - | 1 | 338 | 131 | ||
BC047058.1 | - | 1 | 269 | 131 | ||
BC064602.1 | - | 5 | 1737 | 131 | ||
BC142986.1 | - | 5 | 1750 | 131 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
