Assay Details
| Gene Symbol: KCND3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005270851.4 | XP_005270908.1 | 8 | 3484 | 122 | |
| XM_006710629.3 | XP_006710692.1 | 8 | 3794 | 122 | ||
| XM_006710630.3 | XP_006710693.1 | 7 | 3427 | 122 | ||
| XM_017001244.1 | XP_016856733.1 | 8 | 3809 | 122 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

KCND3
Genomic Map