Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928106 | LOC101928106 | -29.0kb | Sense | Chr. 6 - 10465999 - 10481946 on NCBI Build GRCh38 | |
| Downstream |
glucosaminyl (N-acetyl) transferase 2, I-branching enzyme (I blood group) |
GCNT2 |
CCAT, CTRCT13, GCNT2C, GCNT5, IGNT, II, NACGT1, NAGCT1, ULG3, bA360O19.2, bA421M1.1 |
6.7kb | Anti-sense | Chr. 6 - 10521283 - 10629368 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map