Assay Details
| Gene Symbol: LOC101929260 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110241.1 | 5 - 6 | 1728 | 106 | ||
| GenBank mRNA | BC014936.1 | - | 5 - 6 | 1728 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101929260
Genomic Map