Assay Details
| Gene Symbol: MINOS1P1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_051980.1 | 1 | 1055 | 70 | ||
| GenBank mRNA | AL049782.1 | - | 1 | 1055 | 70 | |
| U50530.1 | - | 1 | 353 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

MINOS1P1
Genomic Map