Assay Details
| Gene Symbol: NCKAP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207363.2 | NP_997246.2 | 14 | 3409 | 120 | |
| XM_005263659.3 | XP_005263716.1 | 13 | 3991 | 120 | ||
| XM_005263660.3 | XP_005263717.1 | 13 | 3506 | 120 | ||
| XM_011511097.2 | XP_011509399.1 | 15 | 4179 | 120 | ||
| XM_011511098.2 | XP_011509400.1 | 15 | 4176 | 120 | ||
| XM_011511099.2 | XP_011509401.1 | 14 | 3574 | 120 | ||
| XM_011511100.2 | XP_011509402.1 | 14 | 3672 | 120 | ||
| XM_011511101.1 | XP_011509403.1 | 12 | 3082 | 120 | ||
| XM_011511102.2 | XP_011509404.1 | 13 | 7508 | 120 | ||
| XM_011511103.2 | XP_011509405.1 | 11 | 3310 | 120 | ||
| XM_011511104.1 | XP_011509406.1 | 11 | 2961 | 120 | ||
| XM_011511105.2 | XP_011509407.1 | 11 | 3752 | 120 | ||
| XM_017003974.1 | XP_016859463.1 | 14 | 3302 | 120 | ||
| XM_017003975.1 | XP_016859464.1 | 13 | 3604 | 120 | ||
| XM_017003976.1 | XP_016859465.1 | 15 | 4200 | 120 | ||
| XM_017003977.1 | XP_016859466.1 | 12 | 4110 | 120 | ||
| XM_017003978.1 | XP_016859467.1 | 8 | 3520 | 120 | ||
| GenBank mRNA | AB005217.1 | - | 1 | 1052 | 120 | |
| AY946008.1 | - | 9 | 2831 | 120 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NCKAP5
Genomic Map