Assay Details
| Gene Symbol: KDM4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146695.1 | NP_001140167.1 | 4 - 5 | 1017 | 62 | |
| NM_001146696.1 | NP_001140168.1 | 4 - 5 | 589 | 62 | ||
| NM_001304339.1 | NP_001291268.1 | 4 - 5 | 1017 | 62 | ||
| NM_001304340.1 | NP_001291269.1 | 3 - 4 | 841 | 62 | ||
| NM_015061.3 | NP_055876.2 | 4 - 5 | 1017 | 62 | ||
| NR_130707.1 | 4 - 5 | 1017 | 62 | |||
| XM_006716741.2 | XP_006716804.1 | 4 - 5 | 686 | 62 | ||
| XM_017014498.1 | XP_016869987.1 | 4 - 5 | 686 | 62 | ||
| XM_017014502.1 | XP_016869991.1 | 4 - 5 | 686 | 62 | ||
| XM_017014503.1 | XP_016869992.1 | 4 - 5 | 686 | 62 | ||
| XM_017014504.1 | XP_016869993.1 | 4 - 5 | 686 | 62 | ||
| XM_017014506.1 | XP_016869995.1 | 4 - 5 | 564 | 62 | ||
| GenBank mRNA | AB018323.1 | - | 4 - 5 | 597 | 62 | |
| AK304032.1 | - | 4 - 5 | 515 | 62 | ||
| AK304095.1 | - | 3 - 4 | 414 | 62 | ||
| AK310439.1 | - | 4 - 5 | 600 | 62 | ||
| AK314355.1 | - | 4 - 5 | 582 | 62 | ||
| BC104859.1 | - | 4 - 5 | 497 | 8143862 | 62 | |
| BC104861.1 | - | 4 - 5 | 497 | 8143864 | 62 | |
| BC143571.1 | - | 4 - 5 | 497 | 9052082 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK304095.1, NM_001304340.1, XM_017014506.1 transcripts.
Set Membership: |
|

KDM4C
Genomic Map