Assay Details
| Gene Symbol: LINC00208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040035.1 | 2 - 3 | 1060 | 82 | ||
| GenBank mRNA | AJ291678.1 | - | 2 - 3 | 1060 | 82 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000088.hg.1
Additional Information:
This assay is in the 5'UTR region of the AJ291678.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05958016 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957990 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05958007 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05958034 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957989 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957985 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957965 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05958005 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957973 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957948 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957998 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16068556 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957994 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05958015 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05958011 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05958013 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957980 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05958025 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26237250 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957949 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26237200 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26237226 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26237230 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16068549 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05958033 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05958028 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957960 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05958032 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957982 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05958027 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957997 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05958020 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26237173 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957975 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05958022 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957970 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16068555 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05958000 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05958030 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957971 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957979 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957974 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26237243 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957967 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957955 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16068551 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26237222 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05958024 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26237191 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05958010 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957978 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05958018 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957954 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05958001 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957951 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26237208 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05958019 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957988 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26237229 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957964 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26237224 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P26237192 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957958 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05958012 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957999 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957957 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05957962 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P82795 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P82795 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 30880 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P130588 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P130588 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 62021 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P82795 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P130588 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P130588 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P82795 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 75942 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 80574 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P18216790 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P32206058 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P130588 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8144641 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8144640 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8144642 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8144639 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000837.hg.1 | TC08000088.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000837.hg.1 | ENST00000304233.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000837.hg.1 | NR_040035.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006749.hg.1 | ENST00000304233.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006749.hg.1 | ENST00000304233.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006749.hg.1 | ENST00000304233.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006749.hg.1 | ENST00000304233.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006749.hg.1 | NR_040035.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006749.hg.1 | NR_040035.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006749.hg.1 | NR_040035.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800145724.hg.1 | ENST00000304233.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145724.hg.1 | NR_040035.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800145724.hg.1 | ENST00000304233.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145724.hg.1 | NR_040035.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800145724.hg.1 | TC0800006749.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800006749.hg.1 | ENST00000304233.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006749.hg.1 | ENST00000304233.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006749.hg.1 | ENST00000304233.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006749.hg.1 | ENST00000304233.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006749.hg.1 | NR_040035.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006749.hg.1 | NR_040035.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006749.hg.1 | NR_040035.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 1552955_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1552955_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A036354 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A036354 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 133077 | - | - |

LINC00208
Genomic Map