Assay Details
| Gene Symbol: FAM167A-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026814.1 | 5 - 6 | 833 | 60 | ||
| GenBank mRNA | AJ301563.1 | - | 5 - 6 | 833 | 60 | |
| BC080558.1 | - | 5 - 6 | 967 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800012267.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012267.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012267.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012267.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012267.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012267.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012267.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012267.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012267.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012267.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012267.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012267.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012267.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012267.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012266.hg.1 | ENST00000529305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012266.hg.1 | ENST00000529305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012266.hg.1 | ENST00000529305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012266.hg.1 | ENST00000529305.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012266.hg.1 | ENST00000624614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012266.hg.1 | ENST00000624614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012266.hg.1 | ENST00000624614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012266.hg.1 | ENST00000624614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012266.hg.1 | ENST00000624614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012266.hg.1 | ENST00000624614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012266.hg.1 | ENST00000624614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012266.hg.1 | ENST00000624614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184576.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184576.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184576.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184576.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184576.hg.1 | TC0800012267.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800184578.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184578.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184578.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184578.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184578.hg.1 | TC0800012267.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800184575.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184575.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184575.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184575.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184575.hg.1 | TC0800012267.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800184579.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184579.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184579.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184579.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184579.hg.1 | TC0800012267.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800012267.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012267.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012267.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012267.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012267.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012267.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012267.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012267.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012267.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012267.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012267.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012267.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012267.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012267.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184574.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184574.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184574.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184574.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184574.hg.1 | TC0800012267.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800184577.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184577.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184577.hg.1 | ENST00000533578.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800184577.hg.1 | NR_026814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800184577.hg.1 | TC0800012267.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800012266.hg.1 | ENST00000529305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012266.hg.1 | ENST00000529305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012266.hg.1 | ENST00000529305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012266.hg.1 | ENST00000529305.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012266.hg.1 | ENST00000624614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012266.hg.1 | ENST00000624614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012266.hg.1 | ENST00000624614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012266.hg.1 | ENST00000624614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012266.hg.1 | ENST00000624614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012266.hg.1 | ENST00000624614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012266.hg.1 | ENST00000624614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012266.hg.1 | ENST00000624614.1 | Ensembl |

FAM167A-AS1
Genomic Map