| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010438.hg.1 |
ENST00000512517.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010438.hg.1 |
ENST00000512517.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010438.hg.1 |
ENST00000512517.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010438.hg.1 |
ENST00000512517.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010438.hg.1 |
ENST00000512517.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010438.hg.1 |
ENST00000512517.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010438.hg.1 |
ENST00000512517.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010438.hg.1 |
ENST00000512517.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010438.hg.1 |
NR_110951.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010438.hg.1 |
NR_110951.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010438.hg.1 |
NR_110951.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010438.hg.1 |
NR_110951.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010438.hg.1 |
NR_110951.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010438.hg.1 |
NR_110951.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010438.hg.1 |
NR_110951.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400010438.hg.1 |
NR_110951.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400171125.hg.1 |
ENST00000512517.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400171125.hg.1 |
NR_110951.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400171125.hg.1 |
ENST00000512517.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400171125.hg.1 |
NR_110951.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400171125.hg.1 |
TC0400010438.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400010438.hg.1 |
ENST00000512517.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010438.hg.1 |
ENST00000512517.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010438.hg.1 |
ENST00000512517.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010438.hg.1 |
ENST00000512517.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010438.hg.1 |
ENST00000512517.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010438.hg.1 |
ENST00000512517.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010438.hg.1 |
ENST00000512517.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010438.hg.1 |
ENST00000512517.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400010438.hg.1 |
NR_110951.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010438.hg.1 |
NR_110951.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010438.hg.1 |
NR_110951.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010438.hg.1 |
NR_110951.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010438.hg.1 |
NR_110951.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010438.hg.1 |
NR_110951.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010438.hg.1 |
NR_110951.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400010438.hg.1 |
NR_110951.1 |
Refseq |