Assay Details
| Gene Symbol: ASTN2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033973.1 | 2 - 3 | 367 | 109 | ||
| GenBank mRNA | AK057447.1 | - | 2 - 3 | 367 | 109 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000595.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09006755.hg.1 | TC09000595.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006755.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006755.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006754.hg.1 | TC09000595.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006754.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006754.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006751.hg.1 | TC09000595.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006751.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006751.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006753.hg.1 | TC09000595.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006753.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006753.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006750.hg.1 | TC09000595.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006750.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006750.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006752.hg.1 | TC09000595.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006752.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09006752.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157215.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157215.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157215.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157215.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157215.hg.1 | TC0900008570.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900157207.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157207.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157207.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157207.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157207.hg.1 | TC0900008570.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900157210.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157210.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157210.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157210.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157210.hg.1 | TC0900008570.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008570.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157211.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157211.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157211.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157211.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157211.hg.1 | TC0900008570.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900157213.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157213.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157213.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157213.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157213.hg.1 | TC0900008570.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900157212.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157212.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157212.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157212.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157212.hg.1 | TC0900008570.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900157216.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157216.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157216.hg.1 | ENST00000418250.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900157216.hg.1 | NR_033973.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900157216.hg.1 | TC0900008570.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A039155 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A039155 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 232552 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3255792 | - | - |

ASTN2-AS1
Genomic Map