Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014488.hg.1 |
ENST00000418006.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014488.hg.1 |
NR_036546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014488.hg.1 |
TC12001093.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014487.hg.1 |
ENST00000418006.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014487.hg.1 |
NR_036546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014487.hg.1 |
TC12001093.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009589.hg.1 |
ENST00000418006.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009589.hg.1 |
ENST00000418006.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009589.hg.1 |
ENST00000418006.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009589.hg.1 |
NR_036546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009589.hg.1 |
NR_036546.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009589.hg.1 |
NR_036546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009589.hg.1 |
ENST00000418006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009589.hg.1 |
ENST00000418006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009589.hg.1 |
ENST00000418006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009589.hg.1 |
NR_036546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009589.hg.1 |
NR_036546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009589.hg.1 |
NR_036546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200171742.hg.1 |
ENST00000418006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171742.hg.1 |
NR_036546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200171742.hg.1 |
ENST00000418006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171742.hg.1 |
NR_036546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200171742.hg.1 |
TC1200009589.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200171744.hg.1 |
ENST00000418006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171744.hg.1 |
NR_036546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200171744.hg.1 |
ENST00000418006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171744.hg.1 |
NR_036546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200171744.hg.1 |
TC1200009589.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200171743.hg.1 |
ENST00000418006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171743.hg.1 |
NR_036546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200171743.hg.1 |
ENST00000418006.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171743.hg.1 |
NR_036546.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200171743.hg.1 |
TC1200009589.hg.1 |
Clariom D TC |