Assay Details
| Gene Symbol: KRTAP5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK057910.1 | - | 1 - 2 | 616 | 123 | |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P33446 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P33446 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 44139 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 60072 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P33446 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P33446 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27995264 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27995313 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599333 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599313 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599307 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27995293 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599304 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599303 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599329 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27995288 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599330 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27995296 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27995281 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599311 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599301 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599306 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16409588 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27995276 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27995270 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599316 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599309 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599302 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599315 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599300 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599317 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599313 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27995313 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27995276 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599307 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27995288 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27995296 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599303 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599309 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27995293 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599317 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16409588 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599333 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27995270 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599311 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599300 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27995264 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599316 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599304 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599301 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599329 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599330 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599315 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599306 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P27995281 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07599302 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P33446 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P33446 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599327 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599306 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599371 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599311 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599354 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16409551 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599348 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599356 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599317 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599326 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599304 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16409546 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599346 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599352 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16409535 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599323 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599337 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16409533 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599300 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599335 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599313 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599307 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599344 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16409534 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599320 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16409537 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599315 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599330 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599372 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599339 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599309 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599336 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16409594 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599341 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16409553 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599333 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599302 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07599301 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_021489_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7945649 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7945650 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7937697 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7945651 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7945648 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7937696 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1562681_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1566219_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000524947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000524947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000524947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000524947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000532922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000532922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000532922.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000534077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | ENST00000534077.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006522.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000424148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000524947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000524947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000524947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000524947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000532922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000532922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000532922.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000534077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | ENST00000534077.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | NR_021489.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006522.hg.1 | NR_021489.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 1562681_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1562681_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1566219_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1566219_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566218_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562681_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566218_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566218_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566218_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562681_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566219_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566219_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566219_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562681_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566219_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566219_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566218_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562681_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566218_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562681_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566218_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562681_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566218_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566219_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562681_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566219_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562681_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566219_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566218_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562681_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562681_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566219_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566219_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566218_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1562681_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566219_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1566218_at8 | - | - |
| Human Genome U95 Set | 57206_r_at | - | - |
| Human Genome U95 Set | 57206_PM_r_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A045958 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A045958 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3248668 | NR_021489.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3248668 | AK055141.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3248668 | NR_021489.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3248668 | AK055141.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3248668 | NR_021489.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3248668 | AK055141.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3248668 | NR_021489.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3248668 | AK055141.1 | human_transcriptome |

KRTAP5-AS1
Genomic Map