Assay Details
| Gene Symbol: DDR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK058006.1 | - | 1 | 1454 | 124 | |
| AK093908.1 | - | 1 | 773 | 124 | ||
| AY871271.1 | - | 1 | 788 | 124 | ||
| BX537651.1 | - | 1 | 2999 | 124 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

DDR2
Genomic Map