| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09012673.hg.1 |
TC09000949.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09012673.hg.1 |
ENST00000380225.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09012673.hg.1 |
NM_002175.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09012671.hg.1 |
TC09000949.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09012671.hg.1 |
ENST00000380225.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09012671.hg.1 |
NM_002175.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009692.hg.1 |
ENST00000380225.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009692.hg.1 |
ENST00000380225.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009692.hg.1 |
ENST00000380225.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009692.hg.1 |
ENST00000380225.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009692.hg.1 |
ENST00000380225.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009692.hg.1 |
NM_002175.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009692.hg.1 |
NM_002175.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009692.hg.1 |
NM_002175.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009692.hg.1 |
NM_002175.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009692.hg.1 |
NM_002175.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166316.hg.1 |
ENST00000380225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166316.hg.1 |
NM_002175.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166316.hg.1 |
ENST00000380225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166316.hg.1 |
NM_002175.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166316.hg.1 |
TC0900009692.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900009692.hg.1 |
ENST00000380225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009692.hg.1 |
ENST00000380225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009692.hg.1 |
ENST00000380225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009692.hg.1 |
ENST00000380225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009692.hg.1 |
ENST00000380225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009692.hg.1 |
NM_002175.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009692.hg.1 |
NM_002175.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009692.hg.1 |
NM_002175.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009692.hg.1 |
NM_002175.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009692.hg.1 |
NM_002175.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166312.hg.1 |
ENST00000380225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166312.hg.1 |
NM_002175.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166312.hg.1 |
ENST00000380225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166312.hg.1 |
NM_002175.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166312.hg.1 |
TC0900009692.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900166313.hg.1 |
ENST00000380225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166313.hg.1 |
NM_002175.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166313.hg.1 |
ENST00000380225.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900166313.hg.1 |
NM_002175.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900166313.hg.1 |
TC0900009692.hg.1 |
Clariom D TC |