Assay Details
| Gene Symbol: OR1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002548.2 | NP_002539.2 | 1 | 654 | 67 | |
| GenBank mRNA | BC069552.1 | - | 1 | 654 | 7262253 | 67 |
| BC106735.1 | - | 1 | 654 | 40015766 | 67 | |
| BC106736.1 | - | 1 | 654 | 40015767 | 67 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein import into nucleus, translocation
movement of cell or subcellular component
chemotaxis
G-protein coupled receptor signaling pathway
single fertilization
sensory perception of smell
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity
protein import into nucleus, translocation
movement of cell or subcellular component
chemotaxis
G-protein coupled receptor signaling pathway
single fertilization
sensory perception of smell
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10196420 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30745808 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30745795 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10196428 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10196443 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30745785 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30745807 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10196440 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30745810 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30745818 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30745788 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30745811 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30745809 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30745787 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30745805 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10196441 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16936239 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30745789 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30745802 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10196430 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10196419 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10196442 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30745784 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10196439 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30745803 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10196421 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 168362 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 47047 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 325973 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P27239 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P27239 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 8950 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P135375 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P135375 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 40757 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P03203421 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P27239 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P27239 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10196420 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745795 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745808 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745812 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16936239 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745787 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745803 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10196430 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10196419 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745786 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745788 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745805 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10196421 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745807 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745810 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745802 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745809 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745811 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745786 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10196420 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745795 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745810 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745787 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745802 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10196419 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745807 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16936239 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745808 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10196421 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10196430 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745788 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745812 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745811 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745805 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745803 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30745809 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P27239 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P135375 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10196431 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10196420 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10196425 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10196421 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10196438 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10196436 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10196433 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10196427 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10196423 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10196429 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10196415 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P10196417 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P135375 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P135375 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P27239 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 154063 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 85438 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P135375 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10196431 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10196427 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10196436 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10196421 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10196438 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10196425 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10196439 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10196417 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10196429 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10196423 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10196440 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10196442 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10196415 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10196433 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10196420 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P135375 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_002548_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8011323 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8011322 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 221464_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11732974_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11732974_at | - | - |
| Human Genome U133 Plus 2.0 Array | 221464_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 221464_at | - | - |
| Human Genome U133A 2.0 Array | 221464_at | - | - |
| Human Genome U133A 2.0 Array | 217224_PM_at | - | - |
| Human Genome U133A 2.0 Array | 221464_PM_at | - | - |
| Human Genome U133A 2.0 Array | 217224_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_002548_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw531216 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221464_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221464_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221464_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221464_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221464_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221464_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221464_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221464_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221464_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221464_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 221464_at4 | - | - |
| Human Genome U133 Set | 221464_PM_at | - | - |
| Human Genome U133 Set | 217224_PM_at | - | - |
| Human Genome U133 Set | 217224_at | - | - |
| Human Genome U133 Set | 221464_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A055007 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A055007 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 118640 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1757546 | BC069552.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1757546 | BC106735.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1757546 | BC106736.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1757546 | NM_002548.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1757546 | BC069552.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1757546 | BC106735.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1757546 | BC106736.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1757546 | NM_002548.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1757546 | BC069552.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1757546 | BC106735.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1757546 | BC106736.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1757546 | NM_002548.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1757546 | BC069552.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1757546 | BC106735.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1757546 | BC106736.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1757546 | NM_002548.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1757546 | BC069552.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1757546 | BC106735.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1757546 | BC106736.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1757546 | NM_002548.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_4505516-S | BC069552.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_4505516-S | BC106735.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_4505516-S | BC106736.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_4505516-S | NM_002548.2 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_19391-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0006420131 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0006420131 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1757546 | BC069552.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1757546 | BC106735.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1757546 | BC106736.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1757546 | NM_002548.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1757546 | BC069552.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1757546 | BC106735.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1757546 | BC106736.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1757546 | NM_002548.2 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_19391 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_19391 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1757546 | BC069552.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1757546 | BC106735.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1757546 | BC106736.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1757546 | NM_002548.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1757546 | BC069552.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1757546 | BC106735.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1757546 | BC106736.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1757546 | NM_002548.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1757546 | BC069552.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1757546 | BC106735.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1757546 | BC106736.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1757546 | NM_002548.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1757546 | BC069552.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1757546 | BC106735.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1757546 | BC106736.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1757546 | NM_002548.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1757546 | BC069552.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1757546 | BC106735.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1757546 | BC106736.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1757546 | NM_002548.2 | human_transcriptome |

OR1D2
Genomic Map