Assay Details
| Gene Symbol: MAPKAPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004759.4 | NP_004750.1 | 7 - 8 | 1104 | 75 | |
| NM_032960.3 | NP_116584.2 | 7 - 8 | 1104 | 75 | ||
| XM_005273353.3 | XP_005273410.1 | 7 - 8 | 1212 | 75 | ||
| XM_017002810.1 | XP_016858299.1 | 7 - 8 | 770 | 75 | ||
| GenBank mRNA | AK307499.1 | - | 7 - 8 | 689 | 75 | |
| BC036060.1 | - | 7 - 8 | 1104 | 5296259 | 75 | |
| BC052584.1 | - | 7 - 8 | 1087 | 6267183 | 75 | |
| X75346.1 | - | 7 - 8 | 882 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001727.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPK activity
toll-like receptor signaling pathway
protein phosphorylation
leukotriene metabolic process
response to stress
inflammatory response
cellular response to DNA damage stimulus
Ras protein signal transduction
peptidyl-serine phosphorylation
G2 DNA damage checkpoint
response to lipopolysaccharide
regulation of interleukin-6 production
regulation of tumor necrosis factor production
response to cytokine
cellular response to vascular endothelial growth factor stimulus
p38MAPK cascade
positive regulation of tumor necrosis factor biosynthetic process
regulation of mRNA stability
macropinocytosis
protein autophosphorylation
vascular endothelial growth factor receptor signaling pathway
inner ear development
3'-UTR-mediated mRNA stabilization
regulation of cellular response to heat
protein kinase activity
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
signal transducer activity
protein binding
calmodulin binding
ATP binding
calcium-dependent protein serine/threonine kinase activity
MAPK cascade
activation of MAPK activity
toll-like receptor signaling pathway
protein phosphorylation
leukotriene metabolic process
response to stress
inflammatory response
cellular response to DNA damage stimulus
Ras protein signal transduction
peptidyl-serine phosphorylation
G2 DNA damage checkpoint
response to lipopolysaccharide
regulation of interleukin-6 production
regulation of tumor necrosis factor production
response to cytokine
cellular response to vascular endothelial growth factor stimulus
p38MAPK cascade
positive regulation of tumor necrosis factor biosynthetic process
regulation of mRNA stability
macropinocytosis
protein autophosphorylation
vascular endothelial growth factor receptor signaling pathway
inner ear development
3'-UTR-mediated mRNA stabilization
regulation of cellular response to heat
protein kinase activity
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
signal transducer activity
protein binding
calmodulin binding
ATP binding
calcium-dependent protein serine/threonine kinase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000479009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000479009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000479009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000479009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000479009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000479009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000479009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000493447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000493447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | ENST00000493447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026946.hg.1 | TC01001727.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026946.hg.1 | ENST00000294981.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026946.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026946.hg.1 | ENST00000479009.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026946.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026946.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183702.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183702.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183702.hg.1 | ENST00000479009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183702.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183702.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183702.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183702.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183702.hg.1 | ENST00000479009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100183702.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183702.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100183702.hg.1 | TC0100011384.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000294981.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000367103.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000479009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000479009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000479009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000479009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000479009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000479009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000479009.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000493447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000493447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | ENST00000493447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_004759.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_032960.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011384.hg.1 | NM_032960.3 | Refseq |

MAPKAPK2
Genomic Map