Assay Details
| Gene Symbol: RAMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005854.2 | NP_005845.2 | 1 - 2 | 167 | 117 | |
| GenBank mRNA | AJ001015.1 | - | 1 - 2 | 117 | ||
| AL548945.3 | - | 1 - 2 | 117 | |||
| AY265458.1 | - | 1 - 2 | 99 | 117 | ||
| BC027975.1 | - | 1 - 2 | 116 | 5229167 | 117 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000534.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
vasculogenesis
response to hypoxia
sprouting angiogenesis
cAMP biosynthetic process
calcium ion transport
intracellular protein transport
G-protein coupled receptor signaling pathway
heart development
female pregnancy
regulation of blood pressure
regulation of G-protein coupled receptor protein signaling pathway
positive regulation of gene expression
protein transport
positive regulation of cAMP biosynthetic process
receptor internalization
response to estradiol
response to progesterone
cellular response to hormone stimulus
adherens junction assembly
cellular response to vascular endothelial growth factor stimulus
negative regulation of vascular permeability
positive regulation of angiogenesis
bicellular tight junction assembly
basement membrane assembly
protein localization to plasma membrane
vascular smooth muscle cell development
negative regulation of endothelial cell apoptotic process
positive regulation of vasculogenesis
protein binding
protein transporter activity
coreceptor activity
angiogenesis
vasculogenesis
response to hypoxia
sprouting angiogenesis
cAMP biosynthetic process
calcium ion transport
intracellular protein transport
G-protein coupled receptor signaling pathway
heart development
female pregnancy
regulation of blood pressure
regulation of G-protein coupled receptor protein signaling pathway
positive regulation of gene expression
protein transport
positive regulation of cAMP biosynthetic process
receptor internalization
response to estradiol
response to progesterone
cellular response to hormone stimulus
adherens junction assembly
cellular response to vascular endothelial growth factor stimulus
negative regulation of vascular permeability
positive regulation of angiogenesis
bicellular tight junction assembly
basement membrane assembly
protein localization to plasma membrane
vascular smooth muscle cell development
negative regulation of endothelial cell apoptotic process
positive regulation of vasculogenesis
protein binding
protein transporter activity
coreceptor activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000253796.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000253796.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000253796.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000253796.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000253796.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000253796.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000253796.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000253796.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000587142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000587142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000587142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000587142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000587142.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000588576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000588576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000588576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000588576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000588576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000588928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000588928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000588928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000588928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000588928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000589683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000589683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000589683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000589683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000589683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000589683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000589683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000589683.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000591972.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | ENST00000591972.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | NM_005854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | NM_005854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | NM_005854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | NM_005854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | NM_005854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | NM_005854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | NM_005854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007913.hg.1 | NM_005854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006494.hg.1 | ENST00000253796.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006494.hg.1 | NM_005854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006494.hg.1 | TC17000534.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006495.hg.1 | ENST00000253796.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006495.hg.1 | NM_005854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006495.hg.1 | TC17000534.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006493.hg.1 | ENST00000253796.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006493.hg.1 | NM_005854.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17006493.hg.1 | TC17000534.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700157681.hg.1 | ENST00000253796.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157681.hg.1 | ENST00000587142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157681.hg.1 | ENST00000588576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157681.hg.1 | ENST00000588928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157681.hg.1 | ENST00000589683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157681.hg.1 | ENST00000591972.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157681.hg.1 | NM_005854.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157681.hg.1 | ENST00000253796.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157681.hg.1 | ENST00000587142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157681.hg.1 | ENST00000588576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157681.hg.1 | ENST00000588928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157681.hg.1 | ENST00000589683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157681.hg.1 | ENST00000591972.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157681.hg.1 | NM_005854.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157681.hg.1 | TC1700007913.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000253796.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000253796.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000253796.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000253796.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000253796.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000253796.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000253796.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000253796.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000587142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000587142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000587142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000587142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000587142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000588576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000588576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000588576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000588576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000588576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000588928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000588928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000588928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000588928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000588928.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000589683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000589683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000589683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000589683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000589683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000589683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000589683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000589683.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000591972.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | ENST00000591972.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | NM_005854.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | NM_005854.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | NM_005854.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | NM_005854.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | NM_005854.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | NM_005854.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | NM_005854.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007913.hg.1 | NM_005854.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157680.hg.1 | ENST00000253796.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157680.hg.1 | ENST00000587142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157680.hg.1 | ENST00000588576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157680.hg.1 | NM_005854.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157680.hg.1 | ENST00000253796.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157680.hg.1 | ENST00000587142.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157680.hg.1 | ENST00000588576.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157680.hg.1 | NM_005854.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157680.hg.1 | TC1700007913.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700157677.hg.1 | ENST00000253796.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157677.hg.1 | NM_005854.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157677.hg.1 | ENST00000253796.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700157677.hg.1 | NM_005854.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700157677.hg.1 | TC1700007913.hg.1 | Clariom D TC |

RAMP2
Genomic Map