Assay Details
| Gene Symbol: TAF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006284.3 | NP_006275.1 | 2 - 3 | 441 | 78 | |
| GenBank mRNA | BC012088.1 | - | 2 - 3 | 398 | 4562773 | 78 |
| BM990572.1 | - | 3 - 4 | 78 | |||
| CR541943.1 | - | 2 - 3 | 386 | 78 | ||
| U13991.1 | - | 2 - 3 | 403 | 78 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001361.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
DNA-templated transcription, initiation
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
histone deubiquitination
histone H3 acetylation
protein homooligomerization
regulation of signal transduction by p53 class mediator
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
histone acetyltransferase activity
protein binding
enzyme binding
estrogen receptor binding
RNA polymerase binding
DNA-templated transcription, initiation
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
histone deubiquitination
histone H3 acetylation
protein homooligomerization
regulation of signal transduction by p53 class mediator
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
histone acetyltransferase activity
protein binding
enzyme binding
estrogen receptor binding
RNA polymerase binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000527248.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000527248.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000527248.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000527248.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000527248.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000531760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000531760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000531760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000531760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000532344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000532344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000532344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000532344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000532344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | ENST00000532344.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013140.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008657.hg.1 | ENST00000514125.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11017120.hg.1 | ENST00000299424.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11017120.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11017120.hg.1 | TC11001361.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100203472.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203472.hg.1 | ENST00000527248.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203472.hg.1 | ENST00000531760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203472.hg.1 | ENST00000532344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203472.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203472.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203472.hg.1 | ENST00000527248.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203472.hg.1 | ENST00000531760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203472.hg.1 | ENST00000532344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203472.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203472.hg.1 | TC1100013140.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400008657.hg.1 | ENST00000514125.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203477.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203477.hg.1 | ENST00000532344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203477.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203477.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203477.hg.1 | ENST00000532344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203477.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203477.hg.1 | TC1100013140.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100203474.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203474.hg.1 | ENST00000531760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203474.hg.1 | ENST00000532344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203474.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203474.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203474.hg.1 | ENST00000531760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203474.hg.1 | ENST00000532344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203474.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203474.hg.1 | TC1100013140.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100203476.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203476.hg.1 | ENST00000527248.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203476.hg.1 | ENST00000532344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203476.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203476.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203476.hg.1 | ENST00000527248.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203476.hg.1 | ENST00000532344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203476.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203476.hg.1 | TC1100013140.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100203480.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203480.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203480.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203480.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203480.hg.1 | TC1100013140.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000527248.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000527248.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000527248.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000527248.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000527248.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000531760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000531760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000531760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000531760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000532344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000532344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000532344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000532344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000532344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | ENST00000532344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013140.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203475.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203475.hg.1 | ENST00000532344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203475.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203475.hg.1 | ENST00000299424.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203475.hg.1 | ENST00000532344.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203475.hg.1 | NM_006284.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203475.hg.1 | TC1100013140.hg.1 | Clariom D TC |

TAF10
Genomic Map