Assay Details
| Gene Symbol: PRDM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF275816.1 | - | 2 - 3 | 360 | 143 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | ENST00000502755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | ENST00000502755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | ENST00000502755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | ENST00000502755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | ENST00000502755.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | ENST00000296682.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | ENST00000502755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | ENST00000502755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | ENST00000502755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | ENST00000502755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | ENST00000502755.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | NM_020227.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500006950.hg.1 | NM_020227.3 | Refseq |

PRDM9
Genomic Map