Assay Details
| Gene Symbol: DRD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000798.4 | NP_000789.1 | 1 | 1092 | 88 | |
| GenBank mRNA | AK313897.1 | - | 1 | 919 | 88 | |
| AY136750.1 | - | 1 | 696 | 88 | ||
| BC009748.2 | - | 1 | 747 | 3928370 | 88 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
synaptic transmission, dopaminergic
response to amphetamine
regulation of systemic arterial blood pressure by vasopressin
norepinephrine-epinephrine vasoconstriction involved in regulation of systemic arterial blood pressure
cellular calcium ion homeostasis
adenylate cyclase-activating G-protein coupled receptor signaling pathway
activation of adenylate cyclase activity
adenylate cyclase-activating dopamine receptor signaling pathway
chemical synaptic transmission
learning
associative learning
positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway
transmission of nerve impulse
negative regulation of NAD(P)H oxidase activity
wound healing
response to cocaine
positive regulation of adenylate cyclase activity
negative regulation of blood pressure
regulation of female receptivity
sensitization
phospholipase C-activating dopamine receptor signaling pathway
long term synaptic depression
cellular response to catecholamine stimulus
reactive oxygen species metabolic process
dopamine neurotransmitter receptor activity, coupled via Gs
dopamine neurotransmitter receptor activity
dopamine binding
synaptic transmission, dopaminergic
response to amphetamine
regulation of systemic arterial blood pressure by vasopressin
norepinephrine-epinephrine vasoconstriction involved in regulation of systemic arterial blood pressure
cellular calcium ion homeostasis
adenylate cyclase-activating G-protein coupled receptor signaling pathway
activation of adenylate cyclase activity
adenylate cyclase-activating dopamine receptor signaling pathway
chemical synaptic transmission
learning
associative learning
positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway
transmission of nerve impulse
negative regulation of NAD(P)H oxidase activity
wound healing
response to cocaine
positive regulation of adenylate cyclase activity
negative regulation of blood pressure
regulation of female receptivity
sensitization
phospholipase C-activating dopamine receptor signaling pathway
long term synaptic depression
cellular response to catecholamine stimulus
reactive oxygen species metabolic process
dopamine neurotransmitter receptor activity, coupled via Gs
dopamine neurotransmitter receptor activity
dopamine binding
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23071781 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23071758 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23071783 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23071830 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02919563 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23071780 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23071772 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02919566 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 594970 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 62040 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 315077 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P862886 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P54274 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0062 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P862886 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0061 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P54274 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 10605 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 36832 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 61612 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 49392 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 26756 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P54274 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P862886 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_32_P54274 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P862886 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15545006 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23071781 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15545047 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02919567 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15544997 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23071780 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02919566 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23071765 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23071783 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02919563 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23071758 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23071772 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23071781 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02919566 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15544997 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02919563 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23071758 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23071783 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23071780 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15545047 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23071772 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23071765 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15545006 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02919567 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_32_P54274 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P862886 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02919564 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02919566 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02919567 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P02919565 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15545035 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P00917158 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P862886 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P54274 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 9177 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 1491 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 2709 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 1499 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P388188 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P54274 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02919565 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15545035 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02919564 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P02919568 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_000798_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8094158 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7905022 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8053727 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7905027 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8053730 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7905024 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7905026 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7905023 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8053729 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8053726 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8094159 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8094157 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8094160 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8053728 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8094156 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8053725 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7905020 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7905025 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 208486_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11732695_at | - | - |
| Affymetrix Human Genome U219 Array | 11732695_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208486_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 208486_at | - | - |
| Human Genome U133A 2.0 Array | 208486_at | - | - |
| Human Genome U133A 2.0 Array | 208486_PM_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_000798_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw596615 | - | - |
| Human Genome U95 Set | 33050_PM_at | - | - |
| Human Genome U95 Set | 33050_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208486_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208486_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208486_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208486_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208486_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208486_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208486_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208486_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208486_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208486_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 208486_at4 | - | - |
| Human Genome U133 Set | 208486_at | - | - |
| Human Genome U133 Set | 208486_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A025282 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A025282 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 131671 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1689043 | BC009748.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1689043 | NM_000798.4 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1689043 | BC009748.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1689043 | NM_000798.4 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1689043 | BC009748.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1689043 | NM_000798.4 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1689043 | BC009748.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1689043 | NM_000798.4 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1689043 | BC009748.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1689043 | NM_000798.4 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_34328907-S | BC009748.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_34328907-S | NM_000798.4 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_27662-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0002120112 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0002120112 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1689043 | BC009748.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1689043 | NM_000798.4 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1689043 | BC009748.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1689043 | NM_000798.4 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 7560053 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_27662 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1689043 | BC009748.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1689043 | NM_000798.4 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1689043 | BC009748.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1689043 | NM_000798.4 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1689043 | BC009748.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1689043 | NM_000798.4 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1689043 | BC009748.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1689043 | NM_000798.4 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1689043 | BC009748.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1689043 | NM_000798.4 | human_transcriptome |

DRD5
Genomic Map