| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Clariom D Assay |
PSR0700147485.hg.1 |
ENST00000328843.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700147485.hg.1 |
ENST00000409508.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700147485.hg.1 |
ENST00000620169.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700147485.hg.1 |
NM_001277115.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700147485.hg.1 |
ENST00000328843.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700147485.hg.1 |
ENST00000409508.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700147485.hg.1 |
ENST00000620169.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700147485.hg.1 |
NM_001277115.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700147485.hg.1 |
TC0700006867.hg.1 |
Clariom D TC |