Assay Details
| Gene Symbol: GBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052941.4 | NP_443173.2 | 5 - 6 | 790 | 77 | |
| GenBank mRNA | AF288814.1 | - | 5 - 6 | 674 | 77 | |
| AK131094.1 | - | 5 - 6 | 703 | 77 | ||
| AL832576.2 | - | 5 - 6 | 720 | 77 | ||
| BC017889.1 | - | 5 - 6 | 765 | 77 | ||
| BC070055.1 | - | 5 - 6 | 727 | 30345441 | 77 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002849.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01044445.hg.1 | TC01002849.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044445.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044445.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | ENST00000471938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | ENST00000471938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | ENST00000481397.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | ENST00000481397.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | ENST00000481397.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | ENST00000481397.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044444.hg.1 | TC01002849.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044444.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044444.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044443.hg.1 | TC01002849.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044443.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044443.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018452.hg.1 | ENST00000294671.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018452.hg.1 | ENST00000294671.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018452.hg.1 | ENST00000294671.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018452.hg.1 | ENST00000294671.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018452.hg.1 | ENST00000294671.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018452.hg.1 | ENST00000294671.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018452.hg.1 | ENST00000294671.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018452.hg.1 | ENST00000294671.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018452.hg.1 | ENST00000294671.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018452.hg.1 | ENST00000294671.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018452.hg.1 | ENST00000294671.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018452.hg.1 | ENST00000294671.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018452.hg.1 | NM_207398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018452.hg.1 | NM_207398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018452.hg.1 | NM_207398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018452.hg.1 | NM_207398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018452.hg.1 | NM_207398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018452.hg.1 | NM_207398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018452.hg.1 | NM_207398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018452.hg.1 | NM_207398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018452.hg.1 | NM_207398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018452.hg.1 | NM_207398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018452.hg.1 | NM_207398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018452.hg.1 | NM_207398.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044442.hg.1 | TC01002849.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044442.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044442.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | ENST00000471938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | ENST00000471938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | ENST00000481397.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | ENST00000481397.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | ENST00000481397.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | ENST00000481397.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014855.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018452.hg.1 | ENST00000294671.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018452.hg.1 | ENST00000294671.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018452.hg.1 | ENST00000294671.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018452.hg.1 | ENST00000294671.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018452.hg.1 | ENST00000294671.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018452.hg.1 | ENST00000294671.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018452.hg.1 | ENST00000294671.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018452.hg.1 | ENST00000294671.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018452.hg.1 | ENST00000294671.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018452.hg.1 | ENST00000294671.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018452.hg.1 | ENST00000294671.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018452.hg.1 | ENST00000294671.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018452.hg.1 | NM_207398.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018452.hg.1 | NM_207398.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018452.hg.1 | NM_207398.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018452.hg.1 | NM_207398.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018452.hg.1 | NM_207398.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018452.hg.1 | NM_207398.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018452.hg.1 | NM_207398.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018452.hg.1 | NM_207398.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018452.hg.1 | NM_207398.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018452.hg.1 | NM_207398.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018452.hg.1 | NM_207398.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018452.hg.1 | NM_207398.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210695.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210695.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210695.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210695.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210695.hg.1 | TC0100014855.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100210696.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210696.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210696.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210696.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210696.hg.1 | TC0100014855.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100210694.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210694.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210694.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210694.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210694.hg.1 | TC0100014855.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100210697.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210697.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210697.hg.1 | ENST00000355754.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210697.hg.1 | NM_052941.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210697.hg.1 | TC0100014855.hg.1 | Clariom D TC |

GBP4
Genomic Map