Assay Details
Gene Symbol: KRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057088.2 | NP_476429.2 | 2 - 3 | 924 | 92 | |
GenBank mRNA | AJ628418.1 | - | 2 - 3 | 813 | 92 | |
AK314987.1 | - | 2 - 3 | 933 | 92 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001535.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12020169.hg.1 | ENST00000309505.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12020169.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12020169.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12020169.hg.1 | TC12001535.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12020166.hg.1 | ENST00000309505.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12020166.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12020166.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12020166.hg.1 | TC12001535.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12020170.hg.1 | ENST00000309505.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12020170.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12020170.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12020170.hg.1 | TC12001535.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200010775.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010775.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010775.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010775.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010775.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010775.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010775.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010775.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010775.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010775.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010775.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010775.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010775.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010775.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010775.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010775.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010775.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010775.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010775.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010775.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010775.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010775.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12020167.hg.1 | ENST00000309505.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12020167.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12020167.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12020167.hg.1 | TC12001535.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR12020168.hg.1 | ENST00000309505.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12020168.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12020168.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12020168.hg.1 | TC12001535.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1200010775.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010775.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010775.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010775.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010775.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010775.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010775.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010775.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010775.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010775.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010775.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010775.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010775.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010775.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010775.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010775.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010775.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010775.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010775.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010775.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010775.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010775.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200182202.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200182202.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200182202.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200182202.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200182202.hg.1 | TC1200010775.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200182201.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200182201.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200182201.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200182201.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200182201.hg.1 | TC1200010775.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200182204.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200182204.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200182204.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200182204.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200182204.hg.1 | TC1200010775.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200182199.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200182199.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200182199.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200182199.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200182199.hg.1 | TC1200010775.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200182200.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200182200.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200182200.hg.1 | ENST00000417996.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200182200.hg.1 | NM_057088.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200182200.hg.1 | TC1200010775.hg.1 | Clariom D TC |
