| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01053344.hg.1 |
TC01003411.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01053344.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01053344.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01053343.hg.1 |
TC01003411.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01053343.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01053343.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
ENST00000490489.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
ENST00000490489.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
ENST00000490489.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
ENST00000494442.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
ENST00000494442.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
ENST00000494442.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
ENST00000494442.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
ENST00000494442.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
ENST00000494442.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016150.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222813.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222813.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222813.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222813.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222813.hg.1 |
TC0100016150.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
ENST00000490489.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
ENST00000490489.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
ENST00000490489.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
ENST00000494442.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
ENST00000494442.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
ENST00000494442.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
ENST00000494442.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
ENST00000494442.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
ENST00000494442.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016150.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222811.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222811.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222811.hg.1 |
ENST00000368029.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100222811.hg.1 |
NM_080878.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100222811.hg.1 |
TC0100016150.hg.1 |
Clariom D TC |