Assay Details
| Gene Symbol: IGFBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000598.4 | NP_000589.2 | 2 - 3 | 761 | 79 | |
| NM_001013398.1 | NP_001013416.1 | 2 - 3 | 779 | 79 | ||
| XM_017012152.1 | XP_016867641.1 | 2 - 3 | 760 | 79 | ||
| GenBank mRNA | AK055632.1 | - | 3 - 4 | 709 | 79 | |
| AK056274.1 | - | 3 - 4 | 633 | 79 | ||
| AK075483.1 | - | 3 - 4 | 674 | 79 | ||
| AK091706.1 | - | 3 - 4 | 504 | 79 | ||
| AK091745.1 | - | 4 - 5 | 574 | 79 | ||
| AK092447.1 | - | 4 - 5 | 589 | 79 | ||
| AK095408.1 | - | 3 - 4 | 648 | 79 | ||
| AK095864.1 | - | 3 - 4 | 435 | 79 | ||
| AK095869.1 | - | 3 - 4 | 605 | 79 | ||
| AK095900.1 | - | 3 - 4 | 617 | 79 | ||
| AK095907.1 | - | 3 - 4 | 627 | 79 | ||
| AK095974.1 | - | 2 - 3 | 455 | 79 | ||
| AK095991.1 | - | 3 - 4 | 605 | 79 | ||
| AK097201.1 | - | 2 - 3 | 469 | 79 | ||
| AK122873.1 | - | 3 - 4 | 692 | 79 | ||
| AK123735.1 | - | 2 - 3 | 403 | 79 | ||
| AK125224.1 | - | 3 - 4 | 470 | 79 | ||
| AK127537.1 | - | 3 - 4 | 589 | 79 | ||
| AK129951.1 | - | 3 - 4 | 360 | 79 | ||
| AK130884.1 | - | 2 - 3 | 273 | 79 | ||
| AK297767.1 | - | 3 - 4 | 716 | 79 | ||
| AK298143.1 | - | 3 - 4 | 626 | 79 | ||
| AK315668.1 | - | 2 - 3 | 761 | 79 | ||
| AL540642.3 | - | 2 - 3 | 752 | 79 | ||
| BC000013.2 | - | 2 - 3 | 727 | 3506666 | 79 | |
| BC018962.2 | - | 2 - 3 | 707 | 3506929 | 79 | |
| BC064987.1 | - | 2 - 3 | 745 | 4850456 | 79 | |
| DQ884396.1 | - | 3 - 4 | 500 | 79 | ||
| DQ884397.1 | - | 3 - 4 | 566 | 79 | ||
| DQ884398.1 | - | 3 - 4 | 545 | 79 | ||
| M31159.1 | - | 2 - 3 | 738 | 79 | ||
| X64875.1 | - | 2 - 3 | 761 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK056274.1, AK075483.1, AK091706.1, AK091745.1, AK095869.1, AK095991.1 transcripts.
Set Membership: |
|

IGFBP3
Genomic Map