Assay Details
| Gene Symbol: RAB1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | M28209.1 | - | 1 - 2 | 70 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the M28209.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000398529.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000398529.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000398529.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000398529.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000398529.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000409751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000409751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000409751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000409751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000409751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000409751.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000409784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000409784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000409784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000409784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000409784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000409784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000409784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000409784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000409784.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000409892.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000409892.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000409892.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000409892.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000409892.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000409892.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000409892.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000494188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000494188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | ENST00000494188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | NM_004161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | NM_004161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | NM_004161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | NM_004161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | NM_004161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | NM_004161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | NM_004161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | NM_004161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | NM_004161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | NM_004161.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | NM_015543.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | NM_015543.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | NM_015543.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | NM_015543.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | NM_015543.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | NM_015543.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | NM_015543.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012854.hg.1 | NM_015543.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015244.hg.1 | ENST00000342609.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015244.hg.1 | ENST00000342609.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000398529.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000398529.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000398529.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000398529.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000398529.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000409751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000409751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000409751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000409751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000409751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000409751.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000409784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000409784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000409784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000409784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000409784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000409784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000409784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000409784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000409784.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000409892.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000409892.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000409892.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000409892.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000409892.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000409892.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000409892.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000494188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000494188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | ENST00000494188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | NM_004161.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | NM_004161.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | NM_004161.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | NM_004161.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | NM_004161.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | NM_004161.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | NM_004161.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | NM_004161.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | NM_004161.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | NM_004161.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | NM_015543.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | NM_015543.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | NM_015543.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | NM_015543.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | NM_015543.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | NM_015543.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | NM_015543.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012854.hg.1 | NM_015543.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015244.hg.1 | ENST00000342609.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015244.hg.1 | ENST00000342609.6 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 122650 | - | - |

RAB1A
Genomic Map