| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009396.hg.1 |
ENST00000312693.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009396.hg.1 |
ENST00000312693.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009396.hg.1 |
ENST00000312693.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009396.hg.1 |
ENST00000312693.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009396.hg.1 |
ENST00000560751.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009396.hg.1 |
ENST00000560751.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009396.hg.1 |
ENST00000560751.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009396.hg.1 |
ENST00000560751.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009396.hg.1 |
ENST00000560751.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009396.hg.1 |
ENST00000560751.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009396.hg.1 |
ENST00000560751.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009396.hg.1 |
NM_005425.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009396.hg.1 |
NM_005425.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009396.hg.1 |
NM_005425.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009396.hg.1 |
ENST00000312693.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009396.hg.1 |
ENST00000312693.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009396.hg.1 |
ENST00000312693.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009396.hg.1 |
ENST00000312693.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009396.hg.1 |
ENST00000560751.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009396.hg.1 |
ENST00000560751.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009396.hg.1 |
ENST00000560751.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009396.hg.1 |
ENST00000560751.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009396.hg.1 |
ENST00000560751.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009396.hg.1 |
ENST00000560751.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009396.hg.1 |
ENST00000560751.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009396.hg.1 |
NM_005425.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009396.hg.1 |
NM_005425.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009396.hg.1 |
NM_005425.4 |
Refseq |