Assay Details
| Gene Symbol: ATP6AP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005765.2 | NP_005756.2 | 1 - 2 | 135 | 120 | |
| GenBank mRNA | AF109363.1 | - | 1 - 2 | 96 | 120 | |
| AF248966.1 | - | 1 - 2 | 140 | 120 | ||
| AF291814.1 | - | 1 - 2 | 122 | 120 | ||
| AK075382.1 | - | 1 - 2 | 193 | 120 | ||
| AK293510.1 | - | 1 - 2 | 117 | 120 | ||
| AK297007.1 | - | 1 - 2 | 106 | 120 | ||
| AK297760.1 | - | 1 - 2 | 205 | 120 | ||
| AK314633.1 | - | 1 - 2 | 86 | 120 | ||
| AK315948.1 | - | 1 - 2 | 66 | 120 | ||
| AY038990.1 | - | 1 - 2 | 138 | 120 | ||
| BC084541.1 | - | 1 - 2 | 115 | 6575732 | 120 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000190.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF109363.1,AF248966.1,AF291814.1,AK075382.1,AK293510.1,AK297007.1,AK297760.1,AK315948.1,AY038990.1,BC084541.1,NM_005765.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002100.hg.1 | TC0X000190.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002100.hg.1 | ENST00000378438.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002100.hg.1 | ENST00000486558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002100.hg.1 | ENST00000535539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002100.hg.1 | ENST00000535777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002100.hg.1 | ENST00000538655.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002100.hg.1 | ENST00000544975.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002100.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002102.hg.1 | TC0X000190.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002102.hg.1 | ENST00000378438.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002102.hg.1 | ENST00000535539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002102.hg.1 | ENST00000535777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002102.hg.1 | ENST00000538655.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X002102.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000479120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000479120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000479120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000487051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000487051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | ENST00000487051.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011075.hg.1 | ENST00000417716.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147676.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147676.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147676.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147676.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147676.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147676.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00147676.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147676.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147676.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147676.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147676.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147676.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00147676.hg.1 | TC0X00007012.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00147675.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147675.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147675.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147675.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00147675.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147675.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147675.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147675.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00147675.hg.1 | TC0X00007012.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00147679.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147679.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147679.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147679.hg.1 | ENST00000487051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147679.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00147679.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147679.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147679.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147679.hg.1 | ENST00000487051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147679.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00147679.hg.1 | TC0X00007012.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100011075.hg.1 | ENST00000417716.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147677.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147677.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147677.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147677.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147677.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147677.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00147677.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147677.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147677.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147677.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147677.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00147677.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00147677.hg.1 | TC0X00007012.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000378438.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000423649.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000436783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000447485.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000479120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000479120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000479120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000486558.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000487051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000487051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | ENST00000487051.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007012.hg.1 | NM_005765.2 | Refseq |

ATP6AP2
Genomic Map