| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009160.hg.1 |
ENST00000449637.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009160.hg.1 |
ENST00000449637.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009160.hg.1 |
ENST00000449637.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009160.hg.1 |
ENST00000449637.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900009160.hg.1 |
ENST00000449637.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007477.hg.1 |
ENST00000306503.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007477.hg.1 |
ENST00000306503.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007477.hg.1 |
ENST00000306503.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007477.hg.1 |
ENST00000306503.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007477.hg.1 |
ENST00000306503.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007477.hg.1 |
ENST00000306503.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007477.hg.1 |
ENST00000394861.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007477.hg.1 |
ENST00000394861.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007477.hg.1 |
ENST00000394861.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007477.hg.1 |
ENST00000394861.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007477.hg.1 |
NM_014011.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007477.hg.1 |
NM_014011.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007477.hg.1 |
NM_014011.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007477.hg.1 |
NM_014011.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007477.hg.1 |
NM_014011.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007477.hg.1 |
NM_144949.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007477.hg.1 |
NM_144949.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007477.hg.1 |
NM_144949.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007477.hg.1 |
NM_144949.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007477.hg.1 |
NM_144949.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900009160.hg.1 |
ENST00000449637.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009160.hg.1 |
ENST00000449637.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009160.hg.1 |
ENST00000449637.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009160.hg.1 |
ENST00000449637.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900009160.hg.1 |
ENST00000449637.1 |
Ensembl |