Assay Details
| Gene Symbol: COMMD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK023070.1 | - | 2 - 3 | 1021 | 60 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000305103.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000305103.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000305103.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000305103.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000305103.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000305103.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000305103.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000402718.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000402718.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000402718.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000402718.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000402718.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000402718.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000402718.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000450361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000450361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000450361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000450361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000450361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000450361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000450361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000450361.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000529143.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000529143.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000529143.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000533270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000533270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000533270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000533270.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000543949.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000543949.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000543949.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000543949.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000543949.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000543949.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000543949.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000543949.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | ENST00000543949.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_001081003.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_001081003.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_001081003.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_001081003.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_001081003.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_001081003.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_001081003.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_001081004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_001081004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_001081004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_001081004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_001081004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_001081004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_001081004.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_001287237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_001287237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_001287237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_001287237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_001287237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_001287237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_001287237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_001287237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_001287237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_014066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_014066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_014066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_014066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_014066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_014066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_014066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012480.hg.1 | NM_014066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000305103.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000305103.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000305103.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000305103.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000305103.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000305103.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000305103.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000402718.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000402718.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000402718.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000402718.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000402718.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000402718.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000402718.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000450361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000450361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000450361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000450361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000450361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000450361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000450361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000450361.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000529143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000529143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000529143.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000533270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000533270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000533270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000533270.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000543949.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000543949.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000543949.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000543949.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000543949.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000543949.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000543949.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000543949.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | ENST00000543949.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_001081003.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_001081003.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_001081003.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_001081003.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_001081003.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_001081003.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_001081003.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_001081004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_001081004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_001081004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_001081004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_001081004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_001081004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_001081004.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_001287237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_001287237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_001287237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_001287237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_001287237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_001287237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_001287237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_001287237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_001287237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_014066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_014066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_014066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_014066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_014066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_014066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_014066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012480.hg.1 | NM_014066.3 | Refseq |

COMMD5
Genomic Map