Assay Details
| Gene Symbol: MFAP3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009554.3 | NP_001009554.1 | 1 - 2 | 119 | 62 | |
| GenBank mRNA | AF327560.1 | - | 1 - 2 | 105 | 62 | |
| AK308263.1 | - | 1 - 2 | 109 | 62 | ||
| BC066912.1 | - | 1 - 2 | 88 | 5268480 | 62 | |
| BM552945.1 | - | 1 - 2 | 131 | 62 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001729.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF327560.1,BC066912.1,NM_001009554.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04023940.hg.1 | TC04001729.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023940.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023940.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023940.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023940.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000502832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000502832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000502832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000502832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000504999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000504999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000504999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000504999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000504999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000504999.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000506764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000506764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000506764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000506764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000506764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000507601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000507601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000507601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000507601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000507601.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000510306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000510306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000510306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000510306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000510306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000512698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000512698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000512698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | ENST00000512698.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023944.hg.1 | TC04001729.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023944.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04023944.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191397.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191397.hg.1 | ENST00000502832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191397.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191397.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191397.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191397.hg.1 | ENST00000502832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400191397.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191397.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400191397.hg.1 | TC0400012992.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000361618.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393702.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000393704.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000502832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000502832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000502832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000502832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000504999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000504999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000504999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000504999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000504999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000504999.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000506764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000506764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000506764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000506764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000506764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000507601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000507601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000507601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000507601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000507601.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000510306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000510306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000510306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000510306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000510306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000512698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000512698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000512698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | ENST00000512698.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001009554.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_001301648.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_021647.7 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012992.hg.1 | NM_021647.7 | Refseq |

MFAP3L
Genomic Map