Assay Details
| Gene Symbol: OTUD7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130901.1 | NP_570971.1 | 6 - 7 | 961 | 80 | |
| GenBank mRNA | AJ430383.1 | - | 6 - 7 | 961 | 80 | |
| BC035668.1 | - | 8 - 9 | 1058 | 80 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001166.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
immune response
protein K11-linked deubiquitination
negative regulation of I-kappaB kinase/NF-kappaB signaling
regulation of innate immune response
regulation of inflammatory response
protein K63-linked deubiquitination
protein K48-linked deubiquitination
protein deubiquitination involved in ubiquitin-dependent protein catabolic process
DNA binding
thiol-dependent ubiquitin-specific protease activity
zinc ion binding
K63-linked polyubiquitin binding
immune response
protein K11-linked deubiquitination
negative regulation of I-kappaB kinase/NF-kappaB signaling
regulation of innate immune response
regulation of inflammatory response
protein K63-linked deubiquitination
protein K48-linked deubiquitination
protein deubiquitination involved in ubiquitin-dependent protein catabolic process
DNA binding
thiol-dependent ubiquitin-specific protease activity
zinc ion binding
K63-linked polyubiquitin binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15010324.hg.1 | ENST00000307050.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15010324.hg.1 | ENST00000382902.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15010324.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15010324.hg.1 | TC15001166.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184062.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184062.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184062.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500184062.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500184062.hg.1 | TC1500010845.hg.1 | Clariom D TC |

OTUD7A
Genomic Map