Assay Details
| Gene Symbol: GPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142466.2 | NP_001135938.1 | 7 - 8 | 984 | 75 | |
| NM_133443.3 | NP_597700.1 | 7 - 8 | 1008 | 75 | ||
| XM_017023790.1 | XP_016879279.1 | 4 - 5 | 481 | 75 | ||
| GenBank mRNA | AK094971.1 | - | 7 - 8 | 908 | 75 | |
| AY029173.2 | - | 7 - 8 | 992 | 75 | ||
| BC062555.1 | - | 7 - 8 | 957 | 6429257 | 75 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000426.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16005893.hg.1 | ENST00000340124.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16005893.hg.1 | ENST00000440783.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16005893.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16005893.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16005893.hg.1 | TC16000426.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600156358.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600156358.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600156358.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600156358.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600156358.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600156358.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600156358.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600156358.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600156358.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600156358.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600156358.hg.1 | TC1600007723.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000340124.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000440783.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562132.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000562801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | ENST00000569193.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_001142466.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007723.hg.1 | NM_133443.3 | Refseq |

GPT2
Genomic Map