| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19004934.hg.1 |
ENST00000303088.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19004934.hg.1 |
ENST00000353145.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19004934.hg.1 |
ENST00000392324.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19004934.hg.1 |
ENST00000407360.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19004934.hg.1 |
ENST00000456252.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19004934.hg.1 |
ENST00000540981.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19004934.hg.1 |
NM_003721.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19004934.hg.1 |
NM_134440.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19004934.hg.1 |
TC19000369.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
ENST00000303088.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
ENST00000392324.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
ENST00000407360.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
ENST00000456252.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
ENST00000535017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
ENST00000540981.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
ENST00000541873.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
ENST00000543118.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
ENST00000545522.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
ENST00000593273.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
NM_001278727.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
NM_001278728.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
NM_003721.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
NM_134440.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
TC1900007425.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
ENST00000303088.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
ENST00000392324.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
ENST00000407360.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
ENST00000456252.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
ENST00000535017.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
ENST00000540981.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
ENST00000541873.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
ENST00000543118.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
ENST00000545522.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
ENST00000593273.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
NM_001278727.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
NM_001278728.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
NM_003721.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900154700.hg.1 |
NM_134440.2 |
Refseq |