Assay Details
| Gene Symbol: MAGEC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138702.1 | NP_619647.1 | 5 - 6 | 1044 | 68 | |
| GenBank mRNA | AF490508.1 | - | 5 - 6 | 1044 | 68 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X000691.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008906.hg.1 | TC0X000691.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008906.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008906.hg.1 | ENST00000448920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008906.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000409007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000409007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000409007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000409007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000409007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000409007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000409007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000409007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000443323.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000443323.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000443323.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000443323.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000443323.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000443323.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000443323.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000443323.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000483584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000483584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000483584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000483584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000483584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000483584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000483584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000483584.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008611.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008904.hg.1 | TC0X000691.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008904.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008904.hg.1 | ENST00000448920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008904.hg.1 | ENST00000483584.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008904.hg.1 | ENST00000544766.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008904.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X008904.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158468.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158468.hg.1 | ENST00000483584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158468.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158468.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158468.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158468.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158468.hg.1 | ENST00000483584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158468.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158468.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158468.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158468.hg.1 | TC0X00008611.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00158469.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158469.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158469.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00158469.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00158469.hg.1 | TC0X00008611.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000298296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000409007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000409007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000409007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000409007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000409007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000409007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000409007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000409007.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000443323.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000443323.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000443323.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000443323.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000443323.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000443323.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000443323.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000443323.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000483584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000483584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000483584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000483584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000483584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000483584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000483584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000483584.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | ENST00000544766.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | NM_138702.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | NM_177456.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008611.hg.1 | NM_177456.2 | Refseq |

MAGEC3
Genomic Map