Assay Details
| Gene Symbol: POP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015918.3 | NP_057002.2 | 2 - 3 | 195 | 71 | |
| XM_011538441.1 | XP_011536743.1 | 2 - 3 | 253 | 71 | ||
| GenBank mRNA | AJ306296.1 | - | 2 - 3 | 182 | 71 | |
| AK223206.1 | - | 2 - 3 | 167 | 71 | ||
| BC012505.1 | - | 2 - 3 | 182 | 4708484 | 71 | |
| BI668578.1 | - | 2 - 3 | 202 | 71 | ||
More Information
Set Membership: |
|

POP5
Genomic Map