Assay Details
| Gene Symbol: PPP4R4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058237.1 | NP_478144.1 | 4 - 5 | 603 | 106 | |
| XM_005267930.2 | XP_005267987.1 | 4 - 5 | 436 | 106 | ||
| XM_011537038.2 | XP_011535340.1 | 4 - 5 | 599 | 106 | ||
| XM_011537039.2 | XP_011535341.1 | 3 - 4 | 344 | 106 | ||
| XM_011537040.2 | XP_011535342.1 | 2 - 3 | 251 | 106 | ||
| XM_017021528.1 | XP_016877017.1 | 2 - 3 | 288 | 106 | ||
| XM_017021529.1 | XP_016877018.1 | 6 - 7 | 958 | 106 | ||
| XM_017021530.1 | XP_016877019.1 | 6 - 7 | 932 | 106 | ||
| GenBank mRNA | AB046842.1 | - | 4 - 5 | 522 | 106 | |
| BC068491.1 | - | 4 - 5 | 562 | 5297335 | 106 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000585.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14007224.hg.1 | ENST00000304338.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007224.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14007224.hg.1 | TC14000585.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400157174.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400157174.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400157174.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400157174.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400157174.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400157174.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400157174.hg.1 | TC1400008058.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000304338.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000328839.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000553661.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000555690.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556470.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | ENST00000556884.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_020958.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008058.hg.1 | NM_058237.1 | Refseq |

PPP4R4
Genomic Map