Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012161.hg.1 |
ENST00000374294.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012161.hg.1 |
ENST00000374294.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012161.hg.1 |
ENST00000374294.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012161.hg.1 |
NM_004125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012161.hg.1 |
NM_004125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012161.hg.1 |
NM_004125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012161.hg.1 |
NM_004125.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006233.hg.1 |
TC09000558.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006233.hg.1 |
ENST00000374293.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006233.hg.1 |
ENST00000374294.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006233.hg.1 |
ENST00000556107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006233.hg.1 |
NM_001017998.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006233.hg.1 |
NM_001198664.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09006233.hg.1 |
NM_004125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184465.hg.1 |
NM_004125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184465.hg.1 |
NM_004125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184465.hg.1 |
TC0900012161.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900184468.hg.1 |
ENST00000374294.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184468.hg.1 |
NM_004125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184468.hg.1 |
ENST00000374294.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184468.hg.1 |
NM_004125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184468.hg.1 |
TC0900012161.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900184467.hg.1 |
ENST00000374294.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184467.hg.1 |
NM_004125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184467.hg.1 |
ENST00000374294.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184467.hg.1 |
NM_004125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184467.hg.1 |
TC0900012161.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900012161.hg.1 |
ENST00000374294.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012161.hg.1 |
ENST00000374294.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012161.hg.1 |
ENST00000374294.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012161.hg.1 |
NM_004125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012161.hg.1 |
NM_004125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012161.hg.1 |
NM_004125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012161.hg.1 |
NM_004125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184466.hg.1 |
ENST00000374294.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184466.hg.1 |
NM_004125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184466.hg.1 |
ENST00000374294.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900184466.hg.1 |
NM_004125.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900184466.hg.1 |
TC0900012161.hg.1 |
Clariom D TC |