Assay Details
| Gene Symbol: MRPL30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145212.3 | NP_660213.1 | 3 - 4 | 324 | 108 | |
| NR_028356.1 | 3 - 4 | 324 | 108 | |||
| GenBank mRNA | AK025547.1 | - | 3 - 4 | 251 | 108 | |
| AK225894.1 | - | 3 - 4 | 220 | 108 | ||
| BC000217.1 | - | 3 - 4 | 214 | 3509919 | 108 | |
| BC022391.1 | - | 3 - 4 | 288 | 4611185 | 108 | |
| BG532984.1 | - | 3 - 4 | 206 | 108 | ||
| BU154006.1 | - | 197 | 108 | |||
| BU533068.1 | - | 3 - 4 | 254 | 108 | ||
| DA310406.1 | - | 3 - 4 | 325 | 108 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

MRPL30
Genomic Map