Assay Details
| Gene Symbol: RBMXL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019610.5 | NP_062556.2 | 1 - 2 | 374 | 65 | |
| GenBank mRNA | AL832554.1 | - | 1 - 2 | 226 | 65 | |
| BC012942.1 | - | 1 - 2 | 33 | 3875565 | 65 | |
| DA390224.1 | - | 1 - 2 | 190 | 65 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002844.hg.1
Additional Information:
This assay is in the 5'UTR region of the AL832554.1,BC012942.1,NM_019610.5 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01044324.hg.1 | TC01002844.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044324.hg.1 | ENST00000260508.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044324.hg.1 | ENST00000399794.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044324.hg.1 | NM_001008661.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044324.hg.1 | NM_001008662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044324.hg.1 | NM_001162536.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044324.hg.1 | NM_019610.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044325.hg.1 | TC01002844.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044325.hg.1 | ENST00000260508.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044325.hg.1 | ENST00000399794.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044325.hg.1 | NM_001008661.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044325.hg.1 | NM_001008662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044325.hg.1 | NM_001162536.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044325.hg.1 | NM_019610.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044326.hg.1 | TC01002844.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044326.hg.1 | ENST00000399794.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044326.hg.1 | NM_001008661.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044326.hg.1 | NM_001008662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044326.hg.1 | NM_001162536.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044326.hg.1 | NM_019610.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044320.hg.1 | TC01002844.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044320.hg.1 | ENST00000260508.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044320.hg.1 | ENST00000321792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044320.hg.1 | ENST00000370491.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044320.hg.1 | ENST00000399794.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044320.hg.1 | ENST00000413769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044320.hg.1 | NM_001008661.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044320.hg.1 | NM_001008662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044320.hg.1 | NM_001162536.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044320.hg.1 | NM_019610.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044315.hg.1 | TC01002844.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044315.hg.1 | ENST00000321792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044315.hg.1 | ENST00000399794.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044315.hg.1 | ENST00000413769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044315.hg.1 | NM_001162536.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044315.hg.1 | NM_019610.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044328.hg.1 | TC01002844.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044328.hg.1 | ENST00000260508.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044328.hg.1 | ENST00000321792.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044328.hg.1 | ENST00000370491.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044328.hg.1 | ENST00000399794.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044328.hg.1 | ENST00000413769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044328.hg.1 | NM_001008661.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044328.hg.1 | NM_001008662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044328.hg.1 | NM_001162536.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01044328.hg.1 | NM_019610.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210557.hg.1 | ENST00000260508.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210557.hg.1 | ENST00000399794.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210557.hg.1 | NM_001008661.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210557.hg.1 | NM_001008662.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210557.hg.1 | NM_001162536.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210557.hg.1 | NM_019610.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210557.hg.1 | ENST00000260508.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210557.hg.1 | ENST00000399794.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210557.hg.1 | NM_001008661.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210557.hg.1 | NM_001008662.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210557.hg.1 | NM_001162536.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210557.hg.1 | NM_019610.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210557.hg.1 | TC0100014848.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100210552.hg.1 | ENST00000260508.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210552.hg.1 | ENST00000321792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210552.hg.1 | ENST00000370486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210552.hg.1 | ENST00000370491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210552.hg.1 | ENST00000399794.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210552.hg.1 | ENST00000413769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210552.hg.1 | NM_001008661.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210552.hg.1 | NM_001008662.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210552.hg.1 | NM_001162536.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210552.hg.1 | NM_019610.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210552.hg.1 | ENST00000260508.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210552.hg.1 | ENST00000321792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210552.hg.1 | ENST00000370486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210552.hg.1 | ENST00000370491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210552.hg.1 | ENST00000399794.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210552.hg.1 | ENST00000413769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210552.hg.1 | NM_001008661.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210552.hg.1 | NM_001008662.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210552.hg.1 | NM_001162536.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210552.hg.1 | NM_019610.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210552.hg.1 | TC0100014848.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100210553.hg.1 | ENST00000260508.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210553.hg.1 | ENST00000321792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210553.hg.1 | ENST00000370486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210553.hg.1 | ENST00000370491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210553.hg.1 | ENST00000399794.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210553.hg.1 | ENST00000413769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210553.hg.1 | NM_001008661.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210553.hg.1 | NM_001008662.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210553.hg.1 | NM_001162536.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210553.hg.1 | NM_019610.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210553.hg.1 | ENST00000260508.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210553.hg.1 | ENST00000321792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210553.hg.1 | ENST00000370486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210553.hg.1 | ENST00000370491.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210553.hg.1 | ENST00000399794.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210553.hg.1 | ENST00000413769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210553.hg.1 | NM_001008661.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210553.hg.1 | NM_001008662.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210553.hg.1 | NM_001162536.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210553.hg.1 | NM_019610.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210553.hg.1 | TC0100014848.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100210559.hg.1 | ENST00000370486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210559.hg.1 | ENST00000399794.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210559.hg.1 | NM_001008661.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210559.hg.1 | NM_001008662.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210559.hg.1 | NM_001162536.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210559.hg.1 | NM_019610.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210559.hg.1 | ENST00000370486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210559.hg.1 | ENST00000399794.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210559.hg.1 | NM_001008661.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210559.hg.1 | NM_001008662.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210559.hg.1 | NM_001162536.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210559.hg.1 | NM_019610.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210559.hg.1 | TC0100014848.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100210548.hg.1 | ENST00000321792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210548.hg.1 | ENST00000399794.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210548.hg.1 | ENST00000413769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210548.hg.1 | NM_001162536.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210548.hg.1 | NM_019610.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210548.hg.1 | ENST00000321792.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210548.hg.1 | ENST00000399794.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210548.hg.1 | ENST00000413769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210548.hg.1 | NM_001162536.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210548.hg.1 | NM_019610.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210548.hg.1 | TC0100014848.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100210558.hg.1 | ENST00000260508.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210558.hg.1 | ENST00000370486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210558.hg.1 | ENST00000399794.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210558.hg.1 | NM_001008661.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210558.hg.1 | NM_001008662.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210558.hg.1 | NM_001162536.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210558.hg.1 | NM_019610.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210558.hg.1 | ENST00000260508.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210558.hg.1 | ENST00000370486.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210558.hg.1 | ENST00000399794.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100210558.hg.1 | NM_001008661.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210558.hg.1 | NM_001008662.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210558.hg.1 | NM_001162536.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210558.hg.1 | NM_019610.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100210558.hg.1 | TC0100014848.hg.1 | Clariom D TC |

RBMXL1
Genomic Map